ENA313839655 Enamine

tert-butyl 4-[2-(methanesulfonyloxy)ethyl]piperidine-1-carboxylate

  • MDL number MFCD11007804
  • Cas Number : 147699-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73605

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73605
Availability details for
Quantity
  • Availability
  • Add to Cart