ENA313839657 Enamine

N-(3-hydroxyphenyl)ethane-1-sulfonamide

  • MDL number MFCD04151192
  • Cas Number : 723757-83-3
  • Molecular Weight : 201.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73608
Availability details for
Quantity
  • Availability
  • Add to Cart