ENA313839664 Enamine

3-[1-(methylsulfanyl)ethyl]aniline

  • MDL number MFCD14608026
  • Cas Number : 1215308-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73621
Availability details for
Quantity
  • Availability
  • Add to Cart