ENA313839666 Enamine

1-phenyl-3-(1,2,3,4-tetrahydroisoquinolin-5-yl)urea

  • MDL number MFCD12719339
  • Cas Number : 1157922-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73630

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73630
Availability details for
Quantity
  • Availability
  • Add to Cart