ENA313839667 Enamine

2-[3-(aminomethyl)phenoxy]acetamide

  • MDL number MFCD09045052
  • Cas Number : 926221-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52133

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52133
Availability details for
Quantity
  • Availability
  • Add to Cart