ENA313839668 Enamine

1-[2-(2-methylphenoxy)ethyl]piperazine dihydrochloride

  • MDL number MFCD09881972
  • Cas Number : 479669-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72621
Availability details for
Quantity
  • Availability
  • Add to Cart