ENA313839669 Enamine

5-benzyl-3-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD10691169
  • Cas Number : 52156-51-1
  • Molecular Weight : 208.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73638
Availability details for
Quantity
  • Availability
  • Add to Cart