ENA313839671 Enamine

N1-methyl-N1-(propan-2-yl)benzene-1,3-diamine

  • MDL number MFCD11132672
  • Cas Number : 1094323-93-9
  • Molecular Weight : 164.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73659
Availability details for
Quantity
  • Availability
  • Add to Cart