ENA313839673 Enamine

N-[2-(4-methoxyphenyl)ethyl]-2-(methylamino)benzamide

  • MDL number MFCD18380630
  • Cas Number : 75652-61-8
  • Molecular Weight : 284.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72622
Availability details for
Quantity
  • Availability
  • Add to Cart