ENA313839687 Enamine

[4-(4-aminophenoxy)phenyl]methanol

  • MDL number MFCD14599905
  • Cas Number : 1269151-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72627
Availability details for
Quantity
  • Availability
  • Add to Cart