ENA313839689 Enamine

1-(2-methoxyphenyl)-1-phenylmethanamine hydrochloride

  • MDL number MFCD08688096
  • Cas Number : 5267-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260-261
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72628
Availability details for
Quantity
  • Availability
  • Add to Cart