ENA313839692 Enamine

3-chloro-6-(methylsulfanyl)pyridazine

  • MDL number MFCD03931476
  • Cas Number : 7145-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72629
Availability details for
Quantity
  • Availability
  • Add to Cart