ENA313839702 Enamine

(4-fluorophenyl)(piperidin-4-yl)methanol

  • MDL number MFCD06809667
  • Cas Number : 54924-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72630
Availability details for
Quantity
  • Availability
  • Add to Cart