ENA313839705 Enamine

1-(4-bromophenyl)-2,2,2-trifluoroethan-1-ol

  • MDL number MFCD12786801
  • Cas Number : 76911-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50...52
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72633

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72633
Availability details for
Quantity
  • Availability
  • Add to Cart