ENA313839709 Enamine

2-[methyl(morpholine-4-sulfonyl)amino]ethan-1-ol

  • MDL number MFCD12772549
  • Cas Number : 1184516-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72640

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72640
Availability details for
Quantity
  • Availability
  • Add to Cart