ENA313839710 Enamine

1-{[3-(aminomethyl)phenyl]methyl}piperidine-3-carboxamide

  • MDL number MFCD09740797
  • Cas Number : 954583-87-0
  • Molecular Weight : 247.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72641
Availability details for
Quantity
  • Availability
  • Add to Cart