ENA321761707 Enamine

ethyl 2-(chloromethyl)-1-ethyl-1H-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD18483125
  • Cas Number : 1311314-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22373
Availability details for
Quantity
  • Availability
  • Add to Cart