ENA321761708 Enamine

2-chloro-N-(7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)acetamide

  • MDL number MFCD18838613
  • Cas Number : 1306603-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24597
Availability details for
Quantity
  • Availability
  • Add to Cart