ENA321761709 Enamine

2-chloro-5-[(4-chlorophenyl)methyl]-4,6-dimethylpyrimidine

  • MDL number MFCD18483126
  • Cas Number : 1311314-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27762
Availability details for
Quantity
  • Availability
  • Add to Cart