ENA321761711 Enamine

ethyl 2-(2-chloropropanamido)-4-methyl-5-(methylcarbamoyl)thiophene-3-carboxylate

  • MDL number MFCD18483127
  • Cas Number : 1311316-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28000
Availability details for
Quantity
  • Availability
  • Add to Cart