ENA321761712 Enamine

2-chloro-N-[1-(naphthalen-2-yl)ethyl]propanamide

  • MDL number MFCD18785428
  • Cas Number : 1311314-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28056
Availability details for
Quantity
  • Availability
  • Add to Cart