ENA321761713 Enamine

2-chloro-N-(3-cyano-4,5-dimethylfuran-2-yl)propanamide

  • MDL number MFCD18838615
  • Cas Number : 1306603-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28097
Availability details for
Quantity
  • Availability
  • Add to Cart