ENA321761715 Enamine

N-[(1-benzyl-3-phenyl-1H-pyrazol-4-yl)methyl]-2-chloroacetamide

  • MDL number MFCD18483128
  • Cas Number : 1311314-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28320

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28320
Availability details for
Quantity
  • Availability
  • Add to Cart