ENA321761717 Enamine

2-chloro-N-(3-phenylprop-2-yn-1-yl)acetamide

  • MDL number MFCD18785429
  • Cas Number : 1311316-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28330
Availability details for
Quantity
  • Availability
  • Add to Cart