ENA321761718 Enamine

2-chloro-1-[2-(3,4-dimethoxyphenyl)pyrrolidin-1-yl]ethan-1-one

  • MDL number MFCD18838617
  • Cas Number : 1306604-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28448
Availability details for
Quantity
  • Availability
  • Add to Cart