ENA321761719 Enamine

(3Z)-3-bromo-4-ethoxy-1,1,1-trifluorobut-3-en-2-one

  • MDL number MFCD06660659
  • Cas Number : 119577-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34744

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34744
Availability details for
Quantity
  • Availability
  • Add to Cart