ENA321761721 Enamine

4-bromo-1-(propan-2-yl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD18838620
  • Cas Number : 1306604-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43727
Availability details for
Quantity
  • Availability
  • Add to Cart