ENA321761722 Enamine

2-chloro-N-[2-(1H-indol-3-yl)-2-phenylethyl]acetamide

  • MDL number MFCD18838621
  • Cas Number : 904627-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-46076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46076
Availability details for
Quantity
  • Availability
  • Add to Cart