ENA321761731 Enamine

2-chloro-1-[4-(piperidine-1-carbonyl)piperidin-1-yl]ethan-1-one

  • MDL number MFCD18838627
  • Cas Number : 1306604-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-46675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46675
Availability details for
Quantity
  • Availability
  • Add to Cart