ENA321761733 Enamine

2-chloro-N-{[4-(methoxymethyl)phenyl]methyl}acetamide

  • MDL number MFCD18838629
  • Cas Number : 1306604-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-46792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46792
Availability details for
Quantity
  • Availability
  • Add to Cart