ENA321761735 Enamine

2-chloro-N-{2-[4-(propan-2-yloxy)phenyl]ethyl}acetamide

  • MDL number MFCD18785431
  • Cas Number : 92105-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-47056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47056
Availability details for
Quantity
  • Availability
  • Add to Cart