ENA321761736 Enamine

2-chloro-1-{4-phenyl-4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}ethan-1-one

  • MDL number MFCD18785432
  • Cas Number : 1311313-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47104
Availability details for
Quantity
  • Availability
  • Add to Cart