ENA321761738 Enamine

2-chloro-N-({2-[(propan-2-yloxy)methyl]phenyl}methyl)acetamide

  • MDL number MFCD18838631
  • Cas Number : 1306606-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-47203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47203
Availability details for
Quantity
  • Availability
  • Add to Cart