ENA321761742 Enamine

2-chloro-N-{[4-(ethoxymethyl)phenyl]methyl}acetamide

  • MDL number MFCD18483134
  • Cas Number : 1311314-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47429
Availability details for
Quantity
  • Availability
  • Add to Cart