ENA321761743 Enamine

2-chloro-N-[1-(5-methyl-1,2,4-oxadiazol-3-yl)cycloheptyl]acetamide

  • MDL number MFCD18838634
  • Cas Number : 1306604-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47488

Safety Information

msds https://www.enaminestore.com/catalog/EN300-47488
Availability details for
Quantity
  • Availability
  • Add to Cart