ENA321761746 Enamine

1-[4-(benzyloxy)piperidin-1-yl]-2-chloroethan-1-one

  • MDL number MFCD18483135
  • Cas Number : 1311316-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47613
Availability details for
Quantity
  • Availability
  • Add to Cart