ENA321761747 Enamine

2-chloro-N-(3,3-dimethylbutyl)acetamide

  • MDL number MFCD18785433
  • Cas Number : 1259915-95-1
  • Molecular Weight : 177.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-47632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47632
Availability details for
Quantity
  • Availability
  • Add to Cart