ENA321761748 Enamine

2-chloro-N-[(3-{[(furan-2-yl)methoxy]methyl}phenyl)methyl]acetamide

  • MDL number MFCD18838636
  • Cas Number : 1306606-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-47667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47667
Availability details for
Quantity
  • Availability
  • Add to Cart