ENA321761750 Enamine

N-(1-tert-butyl-3-methyl-1H-pyrazol-5-yl)-2-chloroacetamide

  • MDL number MFCD18785435
  • Cas Number : 1311313-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-47781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47781
Availability details for
Quantity
  • Availability
  • Add to Cart