ENA321761751 Enamine

2-chloro-N-({3-[(2-ethoxyethoxy)methyl]phenyl}methyl)acetamide

  • MDL number MFCD18838637
  • Cas Number : 1306605-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-47827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47827
Availability details for
Quantity
  • Availability
  • Add to Cart