ENA321761754 Enamine

1-(2-chloroacetyl)-N-methyl-N-(propan-2-yl)piperidine-4-carboxamide

  • MDL number MFCD18483136
  • Cas Number : 1311314-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-47990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47990
Availability details for
Quantity
  • Availability
  • Add to Cart