ENA321761755 Enamine

2-chloro-N-[1-(2-ethoxyphenyl)propyl]acetamide

  • MDL number MFCD18838639
  • Cas Number : 1306604-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-47997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47997
Availability details for
Quantity
  • Availability
  • Add to Cart