ENA321761757 Enamine

2-chloro-N-[cyclopentyl(4-fluorophenyl)methyl]acetamide

  • MDL number MFCD18483138
  • Cas Number : 1311317-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48118
Availability details for
Quantity
  • Availability
  • Add to Cart