ENA321761760 Enamine

2-chloro-N-{[4-fluoro-2-(trifluoromethyl)phenyl]methyl}acetamide

  • MDL number MFCD18785437
  • Cas Number : 1311314-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48176

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48176
Availability details for
Quantity
  • Availability
  • Add to Cart