ENA321761762 Enamine

2-chloro-N-{[3-(propan-2-yloxy)phenyl]methyl}acetamide

  • MDL number MFCD18483140
  • Cas Number : 1311314-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48217

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48217
Availability details for
Quantity
  • Availability
  • Add to Cart