ENA321761763 Enamine

2-chloro-N-[cyclopentyl(phenyl)methyl]acetamide

  • MDL number MFCD18483141
  • Cas Number : 23459-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48241

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48241
Availability details for
Quantity
  • Availability
  • Add to Cart