ENA321761767 Enamine

2-chloro-N-{3-[N-(propan-2-yl)methanesulfonamido]propyl}acetamide

  • MDL number MFCD18917232
  • Cas Number : 1311317-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-48406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48406
Availability details for
Quantity
  • Availability
  • Add to Cart