ENA321761769 Enamine

2-chloro-N-({4-[(propan-2-yloxy)methyl]phenyl}methyl)acetamide

  • MDL number MFCD18483143
  • Cas Number : 1311314-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48425
Availability details for
Quantity
  • Availability
  • Add to Cart