ENA321761770 Enamine

2-chloro-N-[1-(3-phenylpropanoyl)piperidin-4-yl]acetamide

  • MDL number MFCD18785441
  • Cas Number : 1311315-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-48445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48445
Availability details for
Quantity
  • Availability
  • Add to Cart