ENA321761774 Enamine

N-[1-(2-chloroacetyl)piperidin-4-yl]-3-phenylpropanamide

  • MDL number MFCD18838642
  • Cas Number : 1306603-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48976

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48976
Availability details for
Quantity
  • Availability
  • Add to Cart